3-(6-Aminopyridin-3-yl)acrylic acid ethyl ester 95%
Catalog # APOSOR912986-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:192.21 g/mol
Storage Temperature:Ambient
CAS Number:227963-57-7
Formula:C₁₀H₁₂N₂O₂
MDL Number:MFCD04039876