Lamotrigine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:84057-84-1
:
Formula:C₉H₇Cl₂N₅
MDL Number:MFCD00865333
:256.09 g/mol
Melting Pt:184 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
Frequently Bought Together