
Methyl-6-bromo-1H-indole-2-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:372089-59-3
:
Formula:C₁₀H₈BrNO₂
MDL Number:MFCD00590200
:254.0800 g/mol
Melting Pt:170…190 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together