Order Entry
Slovenia
ContactUsLinkComponent
5-Iodo-1-methyl-1H-pyrazole ≥98%
5-Iodo-1-methyl-1H-pyrazole ≥98%
Catalog # APOSOR903445-25G
Supplier:  Apollo Scientific
CAS Number:  
5-Iodo-1-methyl-1H-pyrazole ≥98%
Catalog # APOSOR903445-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :208.00 g/mol
Storage Temperature:Ambient
CAS Number:34091-51-5
Formula:C₄H₅IN₂
MDL Number:MFCD11109327