Order Entry
Slovenia
ContactUsLinkComponent
Methyl tetrahydropyran-4-ylacetate 96%
Methyl tetrahydropyran-4-ylacetate 96%
Catalog # APOSOR900002-5G
Supplier:  Apollo Scientific
CAS Number:  
Methyl tetrahydropyran-4-ylacetate 96%
Catalog # APOSOR900002-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :158.2000 g/mol
Storage Temperature:Ambient
CAS Number:156002-64-1
Formula:C₈H₁₄O₃
MDL Number:MFCD09972087