2-Chloro-4-pyridineboronic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:458532-96-2
:
Formula:C₅H₅BClNO₂
MDL Number:MFCD03788416
:157.36 g/mol
Melting Pt:155…164 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together