Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(4-Chlorophenyl)piperidine
4-(4-Chlorophenyl)piperidine 95%
4-(4-Chlorophenyl)piperidine 95%
APOSOR8735-1G
:
Apollo Scientific
:
4-(4-Chlorophenyl)piperidine 95%
APOSOR8735-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
195.69 g/mol
:
Ambient
:
26905-02-2
:
C₁₁H₁₄ClN
:
MFCD03077010