
(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:18881-17-9
:
Formula:C₁₀H₁₃NO
MDL Number:MFCD01631316
:163.22 g/mol
Melting Pt:114…116 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together