
4-Piperidone monohydrate hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:40064-34-4
:320589-77-3
Formula:C₅H₉NO·xHCl·yH₂O
MDL Number:MFCD00012776
Melting Pt:94…96 °C
:99.13 g/mol (free base)
EINECS:254-779-9
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together