Order Entry
Slovenia
ContactUsLinkComponent
2,5-Dibromothiophene-3-carboxylic acid 97%
2,5-Dibromothiophene-3-carboxylic acid 97%
Catalog # APOSOR6629-25G
Supplier:  Apollo Scientific
CAS Number:  
2,5-Dibromothiophene-3-carboxylic acid 97%
Catalog # APOSOR6629-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :285.94 g/mol
Storage Temperature:Refrigerator
CAS Number:7311-70-8
Formula:C₅H₂Br₂O₂S
MDL Number:MFCD08445656