Order Entry
Slovenia
ContactUsLinkComponent
2-Mercapto-1H-benzimidazole-5-carboxylic acid 97%
2-Mercapto-1H-benzimidazole-5-carboxylic acid 97%
Catalog # APOSOR6308-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Mercapto-1H-benzimidazole-5-carboxylic acid 97%
Catalog # APOSOR6308-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :194.21 g/mol
Storage Temperature:Ambient
CAS Number:58089-25-1
Formula:C₈H₆N₂O₂S
MDL Number:MFCD01318607