Order Entry
Slovenia
ContactUsLinkComponent
4-Amino-3-bromobenzoic acid 98%
4-Amino-3-bromobenzoic acid 98%
Catalog # APOSOR6241-25G
Supplier:  Apollo Scientific
CAS Number:  
4-Amino-3-bromobenzoic acid 98%
Catalog # APOSOR6241-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :216.0320 g/mol
Melting Pt:211…215 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
CAS Number:6311-37-1
Formula:C₇H₆BrNO₂
MDL Number:MFCD03407439