
4-Indoleboronic acid pinacol ester
Catalog # APOSOR60010-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:243.11 g/mol
Melting Pt:168…173 °C
:GHS07
Storage Temperature:Ambient
CAS Number:388116-27-6
Formula:C₁₄H₁₈BNO₂
MDL Number:MFCD08689896