
Potassium thiocyanate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:333-20-0
:
Formula:KSCN
MDL Number:MFCD00011413
Boiling Pt:500 °C
Melting Pt:172…175 °C
Density:1.89 g/cm³ (25 °C)
:97.18 g/mol
Storage Temperature:Ambient
EINECS:206-370-1
:GHS07
Frequently Bought Together