
(1R,3S)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid 97+%
Catalog # APOSOR52690-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:243.30 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:222530-39-4
Formula:C₁₂H₂₁NO₄
MDL Number:MFCD04973107