Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-3-nitrobenzoic acid
4-Chloro-3-nitrobenzoic acid 98%
4-Chloro-3-nitrobenzoic acid 98%
APOSOR51729-100G
:
Apollo Scientific
:
4-Chloro-3-nitrobenzoic acid 98%
APOSOR51729-100G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 g
Seq:
3401
Environmentally Preferable:
:
:
201.57 g/mol
:
GHS07
:
Ambient
:
96-99-1
:
C₇H₄ClNO₄
:
MFCD00007079
:
180…183 °C