2-Amino-3-bromo-6-chloropyrazine
Catalog # APOSOR480567-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:208.4400 g/mol
Storage Temperature:Ambient
CAS Number:212779-21-0
Formula:C₄H₃BrClN₃
MDL Number:MFCD11044158