Order Entry
Slovenia
ContactUsLinkComponent
Isoquinoline-5-carbonitrile
Irritant
Isoquinoline-5-carbonitrile
Catalog # APOSOR48050-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Isoquinoline-5-carbonitrile
Catalog # APOSOR48050-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :154.1700 g/mol
Melting Pt:135…139 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:27655-41-0
Formula:C₁₀H₆N₂
MDL Number:MFCD03109890