0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
L-Threonine benzyl ester hydrochloride
L-Threonine benzyl ester hydrochloride 98%
L-Threonine benzyl ester hydrochloride 98%
APOSOR471632-25G
:
Apollo Scientific
:
L-Threonine benzyl ester hydrochloride 98%
APOSOR471632-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
245.70 g/mol
:
Ambient
:
33645-24-8
:
C₁₁H₁₅NO₃
:
MFCD00038977