4-Bromo-3,5-dimethyl-1H-pyrrole-2-carboxylic acid
Catalog # APOSOR46656-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:218.06 g/mol
Storage Temperature:Ambient
CAS Number:562074-46-8
Formula:C₇H₈BrNO₂
MDL Number:MFCD11557240