2,3-Dimethyl-1-butene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:563-78-0
:
Formula:C₆H₁₂
MDL Number:MFCD00008923
Boiling Pt:56 °C
Melting Pt:-157
Density:0.68 g/cm³ (25 °C)
Molecular Weight:84.16 g/mol
Flash Pt:–18 °C
Storage Temperature:Ambient
EINECS:209-262-2
UN:3295
ADR:3 II
:GHS02
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together