
2-Chloro-6-methylnicotinic acid methyl ester
Catalog # APOSOR46529-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:185.61 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:53277-47-7
Formula:C₈H₈ClNO₂
MDL Number:MFCD11845763