
Methyl-2-chloropyrimidine-4-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:149849-94-5
:
Formula:C₆H₅ClN₂O₂
MDL Number:MFCD11841080
:172.5691 g/mol
Melting Pt:97…98.5 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together