Order Entry
Slovenia
ContactUsLinkComponent
2,6-Dimethoxyphenylboronic acid
2,6-Dimethoxyphenylboronic acid
Catalog # APOSOR4443-100G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Dimethoxyphenylboronic acid
Catalog # APOSOR4443-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:

Specifications

About this item

 :
 :181.98 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:23112-96-1
Formula:C₈H₁₁BO₄
MDL Number:MFCD01318987
Melting Pt:110…112 °C