Order Entry
Slovenia
ContactUsLinkComponent
2,4-Bis(benzyloxy)-5-bromopyrimidine 96%
2,4-Bis(benzyloxy)-5-bromopyrimidine 96%
Catalog # APOSOR4413-5G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Bis(benzyloxy)-5-bromopyrimidine 96%
Catalog # APOSOR4413-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :371.2300 g/mol
Storage Temperature:Ambient
CAS Number:41244-53-5
Formula:C₁₈H₁₅BrN₂O₂
MDL Number:MFCD00267932