2,5-Dichloropyridine-6-carboxaldehyde
Catalog # APOSOR40362-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:176 g/mol
Melting Pt:105…107 °C
:GHS07
Storage Temperature:Ambient
CAS Number:343781-53-3
Formula:C₆H₃Cl₂NO
MDL Number:MFCD12963768