
Methyl-2-bromo-5-ethylthiazole-4-carboxylate
Catalog # APOSOR40280-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:250.12 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:81569-46-2
Formula:C₇H₈BrNO₂S
MDL Number:MFCD11706992