Order Entry
Slovenia
ContactUsLinkComponent
Methyl-2-bromo-5-chlorothiazole-4-carboxylate 98%
Irritant
Methyl-2-bromo-5-chlorothiazole-4-carboxylate 98%
Catalog # APOSOR40279-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-2-bromo-5-chlorothiazole-4-carboxylate 98%
Catalog # APOSOR40279-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :256.51 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1053655-63-2
Formula:C₅H₃BrClNO₂S
MDL Number:MFCD08704624