2-Iodo-1,3-dimethoxybenzene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16932-44-8
:
Formula:C₈H₉IO₂
MDL Number:MFCD05865214
:264.0603 g/mol
Melting Pt:105…106 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
Frequently Bought Together