
Methyl-2-bromo-5-propylthiazole-4-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1120214-96-1
:
Formula:C₈H₁₀BrNO₂S
MDL Number:MFCD11706993
:264.15 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together