Order Entry
Slovenia
ContactUsLinkComponent
4-Methoxy-3,5-dimethylbenzaldehyde
4-Methoxy-3,5-dimethylbenzaldehyde
Catalog # APOSOR400287-5G
Supplier:  Apollo Scientific
CAS Number:  
4-Methoxy-3,5-dimethylbenzaldehyde
Catalog # APOSOR400287-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :164.21 g/mol
Storage Temperature:Ambient
CAS Number:39250-90-3
Formula:C₁₀H₁₂O₂
MDL Number:MFCD02261755