
4-Oxoheptanedioc acid diethyl ester 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6317-49-3
:
Formula:C₁₁H₁₈O₅
MDL Number:MFCD00009210
:230.26 g/mol
Boiling Pt:144 °C/0.6mmHg
Density:1.073 g/cm³ (25 °C)
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together