Order Entry
Slovenia
ContactUsLinkComponent
5-Methylisatoic anhydride
5-Methylisatoic anhydride
Catalog # APOSOR400012-5G
Supplier:  Apollo Scientific
CAS Number:  
5-Methylisatoic anhydride
Catalog # APOSOR400012-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :177.1568 g/mol
Storage Temperature:Ambient
CAS Number:4692-99-3
Formula:C₉H₇NO₃
MDL Number:MFCD06809783