
7-Chloro-1H-indazole
Catalog # APOSOR3680-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:152.5810 g/mol
Melting Pt:135…137 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
CAS Number:37435-12-4
Formula:C₇H₅ClN₂
MDL Number:MFCD07368214