Order Entry
Slovenia
ContactUsLinkComponent
1-Cyclopentenylboronic acid pinacol ester
Irritant
1-Cyclopentenylboronic acid pinacol ester
Catalog # APOSOR360110-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Cyclopentenylboronic acid pinacol ester
Catalog # APOSOR360110-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :194.0784 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:287944-10-9
Formula:C₁₁H₁₉BO₂
MDL Number:MFCD08669391