4-Phenyl-1H-pyrrole-3-carboxylic acid 95+%
Catalog # APOSOR346623-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:187.2 g/mol
Storage Temperature:Ambient
CAS Number:132040-12-1
Formula:C₁₁H₉NO₂
MDL Number:MFCD06411253