4H-Thieno[3,2-b]pyrrole-5-carboxylic acid hydrazide
Catalog # APOSOR346266-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:181.22 g/mol
Storage Temperature:Ambient
CAS Number:119448-43-0
Formula:C₇H₇N₃OS
MDL Number:MFCD04208027