(1S,3R)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid
Catalog # APOSOR321497-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:243.30 g/mol
Storage Temperature:Refrigerator
CAS Number:222530-34-9
Formula:C₁₂H₂₁NO₄
MDL Number:MFCD06227743