Order Entry
Slovenia
ContactUsLinkComponent
2,5-Pyrazinedicarboxylic acid dihydrate 95%
Irritant
2,5-Pyrazinedicarboxylic acid dihydrate 95%
Catalog # APOSOR315199-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,5-Pyrazinedicarboxylic acid dihydrate 95%
Catalog # APOSOR315199-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :186.123 g/mol
Melting Pt:272…277 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:205692-63-3
Formula:C₆H₆N₂O₅
MDL Number:MFCD00216919