
(R)-tert-Butyl methyl(pyrrolidin-3-yl)carbamate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:392338-15-7
:
Formula:C₁₀H₂₀N₂O₂
MDL Number:MFCD09263387
:200.2800 g/mol
Boiling Pt:268 °C
Melting Pt:30…35 °C
Density:1.03 g/cm³ (25 °C)
Flash Pt:116 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together