
2-Methyl-2H-indazole-5-boronic acid pinacol ester
Catalog # APOSOR30597-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:258.1239 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1189746-27-7
Formula:C₁₄H₁₉BN₂O₂
MDL Number:MFCD11109436