5-Iodopyrimidine
Catalog # APOSOR305385-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:205.9845 g/mol
Melting Pt:126 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:31462-58-5
Formula:C4H3IN2
MDL Number:MFCD00233961