9,9-Di-n-octylfluorene-2-boronic acid pinacol ester
Catalog # APOSOR303500-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:516.61 g/mol
Melting Pt:50…56 °C
:GHS07
Storage Temperature:Ambient
CAS Number:302554-81-0
:MFCD20926226
Formula:C₃₅H₅₃BO₂
MDL Number:MFCD16294553