Order Entry
Slovenia
ContactUsLinkComponent
Methyl-3-methyl-4-nitrobenzoate 98%
Irritant
Methyl-3-methyl-4-nitrobenzoate 98%
Catalog # APOSOR30249-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-3-methyl-4-nitrobenzoate 98%
Catalog # APOSOR30249-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Melting Pt:80…82 °C
 :195.18 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:24078-21-5
Formula:C₉H₉NO₄
MDL Number:MFCD00085640