6-Benzyl-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidine-2,4(1H,3H)-dione 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:135481-57-1
:
Formula:C₁₄H₁₅N₃O₂
MDL Number:MFCD06657750
:257.29 g/mol
Storage Temperature:Ambient
Frequently Bought Together