Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

2-Furonitrile 98%

New Product
Supplier:   Apollo Scientific
2-Furonitrile 98%
Supplier:  Apollo Scientific
New Product
Certificates

5 Options Available Below

Product Details & Documents
Synonyms:  2-Cyanofuran
  • CAS Number:617-90-3
  • Formula:C₅H₃NO
  • MDL Number:MFCD00003223
  • Molecular Weight:93.08 g/mol
  • Boiling Pt:146…148 °C
  • Density:1.064 g/cm³ (25 °C)
  • Flash Pt:35 °C
  •  :Density: 1.085 g/cm³ (20 °C)
  • Storage Temperature:Ambient
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    APOSOR30131-1G
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    APOSOR30131-5G
    New Product
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 105,00 €
    105,00 €
    1 * 25 g
       105,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSOR30131-25G
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 310,00 €
    310,00 €
    1 * 100 g
       310,00 €

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    APOSOR30131-100G
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 540,00 €
    1 540,00 €
    1 * 500 g
       1 540,00 €

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    APOSOR30131-500G
    New Product