3-(tert-Butoxycarbonylethoxycarbonylmethylamino)propionic acid ethyl ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:146256-97-5
:
Formula:C₁₄H₂₅NO₆
MDL Number:MFCD15142792
:303.36 g/mol
Storage Temperature:Ambient
Frequently Bought Together