Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Cyclohexanecarbonitrile

Cyclohexanecarbonitrile

Toxic
Supplier:   Apollo Scientific
Cyclohexanecarbonitrile
Supplier:  Apollo Scientific
Certificates
Cyclohexanecarbonitrile

2 Options Available Below

Product Details & Documents
  • CAS Number:766-05-2
  • Formula:C₇H₁₁N
  • MDL Number:MFCD00040444
  • Molecular Weight:109.17 g/mol
  • Boiling Pt:75…76 °C/16mmHg
  • Melting Pt:11 °C
  • Density:0.919 g/cm³ (25 °C)
  • Flash Pt:65 °C
  • Storage Temperature:Ambient
  • EINECS:212-157-4
  • UN:3276
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 29,00 €
    29,00 €
    1 * 5 g
       29,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.