Order Entry
Slovenia
ContactUsLinkComponent
 
Irritant
cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid
Catalog # APOSOR2627-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid
Catalog # APOSOR2627-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :229.276 g/mol
Melting Pt:90…94 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:136315-70-3
Formula:C₁₁H₁₉NO₄
MDL Number:MFCD00800539