
3-Chloro-2,6-dimethoxy-5-nitrobenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175135-56-5
:
Formula:C₉H₈ClNO₆
MDL Number:MFCD00052955
:261.62 g/mol
Melting Pt:129…132 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together